(3R,4R)-1-Benzylpyrrolidine-3,4-diol

Lieferant: Apollo Scientific
Achtung

Synonyme: (3R,4R)-(-)-1-Benzyl-3,4-pyrrolidindiol

OR322563-5G
APOSOR322563-5GEA 206 EUR
APOSOR322563-5G
(3R,4R)-1-Benzylpyrrolidine-3,4-diol
(3R,4R)-1-Benzylpyrrolidine-3,4-diol
Formel: C₁₁H₁₅NO₂
Molecular Weight: 193.2423 g/mol
Schmelzpunkt: 94…100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01073894
CAS-Nummer: 163439-82-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR