2,2-Dimethyl-7-propoxy-3,4-dihydro-2H-1-benzopyran-4-one, tech

Lieferant: Apollo Scientific

OR32257-1G
APOSOR32257-1GEA 1570 EUR
APOSOR32257-1G
2,2-Dimethyl-7-propoxy-3,4-dihydro-2H-1-benzopyran-4-one, tech
2,2-Dimethyl-7-propoxy-3,4-dihydro-2H-1-benzopyran-4-one
Formel: C₁₄H₁₈O₃
Molecular Weight: 234,291 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03617868
CAS-Nummer: 115613-80-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR