Boc-L-Serin-Methylester

Lieferant: Apollo Scientific

Synonyme: Boc-Ser-OMe, N-Boc-L-serine Methyl Ester, BOC-L-Serine methyl ester

OR322570-500G
APOSOR322570-500GEA 202 EUR
APOSOR322570-500G
Boc-L-Serin-Methylester
Boc-L-Serin-Methylester
Formel: C₉H₁₇NO₅
Dichte: 1,082 g/cm³ (25 °C)
Flammpunkt: 113 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00191869
CAS-Nummer: 2766-43-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR