Bis(2,4-dimethoxybenzyl)amin
Lieferant: Apollo Scientific
OR322572-1G
APOSOR322572-1GEA
53.8
EUR
APOSOR322572-1G
Bis(2,4-dimethoxybenzyl)amin
Bis(2,4-dimethoxybenzyl)amin
Formel:
C₁₈H₂₃NO₄ Molecular Weight: 317,3800 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040480 CAS-Nummer: 20781-23-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...