Pyrido[2,3-d]pyrimidine-2,4(1h,3h)-dione

Lieferant: Apollo Scientific

Synonyme: Pyrido[2,3-d]pyrimidine-2,4(1H,3H)-dione

OR322574-1G
APOSOR322574-1GEA 72.5 EUR
APOSOR322574-1G
Pyrido[2,3-d]pyrimidine-2,4(1h,3h)-dione
Pyrido[2,3-d]pyrimidine-2,4(1h,3h)-dione
Formel: C₇H₅N₃O₂
Molecular Weight: 163,134 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08234704
CAS-Nummer: 21038-66-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR