3-Iod-1H-pyrazol

Lieferant: Apollo Scientific

Synonyme: 3-Iodopyrazole

OR322575-1G
APOSOR322575-1GEA 53.8 EUR
APOSOR322575-1G
3-Iod-1H-pyrazol
3-Iodo-1H-pyrazole
Formel: C₃H₃IN₂
Molecular Weight: 193.98 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12022324
CAS-Nummer: 4522-35-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR