2-(2-Hydroxyethyl)-3,4-dihydro-2H-1,4-benzoxazin-3-one, tech
Lieferant: Apollo Scientific
OR32258-1G
APOSOR32258-1GEA
1570
EUR
APOSOR32258-1G
2-(2-Hydroxyethyl)-3,4-dihydro-2H-1,4-benzoxazin-3-one, tech
2-(2-Hydroxyethyl)-3,4-dihydro-2H-1,4-benzoxazin-3-one
Formel:
C₁₀H₁₁NO₃ Molecular Weight: 193.199 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03617589 CAS-Nummer: 161176-99-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...