N-(3-Bromophenyl)methanesulphonamide

Lieferant: Apollo Scientific

OR322586-1G
APOSOR322586-1GEA 88.6 EUR
APOSOR322586-1G
N-(3-Bromophenyl)methanesulphonamide
N-(3-Bromophenyl)methanesulphonamide
Formel: C₇H₈BrNO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD01213915
CAS-Nummer: 83922-51-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR