5,7-Dimethylisatin 97%

Lieferant: Apollo Scientific

Synonyme: 5,7-Dimethylindolin-2,3-dion, 5,7-Dimethyl-2,3-indolindion, 5,7-Dimethyl-2,3-dioxoindolin, 5,7-Dimethyl-1H-indol-2,3-dion

OR322609-25G
APOSOR322609-25GEA 68.3 EUR
APOSOR322609-25G
5,7-Dimethylisatin 97%
5,7-Dimethylisatin
Formel: C₁₀H₉NO₂
Molecular Weight: 175,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00047219
CAS-Nummer: 39603-24-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR