N-Methylbenzohydrazide

Lieferant: Apollo Scientific

OR322621-250MG
APOSOR322621-250MGEA 104 EUR
APOSOR322621-250MG
N-Methylbenzohydrazide
N-Methylbenzohydrazide
Formel: C₈H₁₀N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00728476
CAS-Nummer: 1483-24-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR