N-(2-Bromethyl)phthalimid 98%

Lieferant: Apollo Scientific

Synonyme: N-(2-Bromethyl)phthalsäureimid, N-(2-Bromethyl)-1,2-benzoldicarbonsäureimid

OR322625-100G
APOSOR322625-100GEA 92.8 EUR
APOSOR322625-100G
N-(2-Bromethyl)phthalimid 98%
N-(2-Bromethyl)phthalimid
Formel: C₁₀H₈BrNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00005902
CAS-Nummer: 574-98-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR