2-(Dimethylamino)thiazole-5-carboxaldehyde 97%
Lieferant: Apollo Scientific
Synonyme:
5-Thiazolecarboxaldehyde, 2-(dimethylamino)-, 2-(dimethylamino)-1,3-thiazole-5-carbaldehyde
OR322627-1G
APOSOR322627-1GEA
306
EUR
APOSOR322627-1G
2-(Dimethylamino)thiazole-5-carboxaldehyde 97%
2-(Dimethylamino)thiazole-5-carboxaldehyde
Formel:
C₆H₈N₂OS Molecular Weight: 156,2 g/mol Schmelzpunkt: 74…77 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03844445 CAS-Nummer: 1005-28-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...