5-Bromobenzo[d]oxazol-2-amine

Lieferant: Apollo Scientific
Achtung

OR322634-25G OR322634-5G
APOSOR322634-25GEA 0 EUR
APOSOR322634-25G APOSOR322634-5G
5-Bromobenzo[d]oxazol-2-amine
5-Bromobenzo[d]oxazol-2-amine
Formel: C₇H₅BrN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD01664213
CAS-Nummer: 64037-07-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR