3-Acetamidophenyl acetate

Lieferant: Apollo Scientific

OR322649-250MG
APOSOR322649-250MGEA 37.7 EUR
APOSOR322649-250MG
3-Acetamidophenyl acetate
3-Acetamidophenyl acetate
Formel: C₁₀H₁₁NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00465580
CAS-Nummer: 6317-89-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR