Ethyl-(S)-(+)-2-pyrrolidinone-5-carboxylate 98%

Lieferant: Apollo Scientific

Synonyme: Ethyl (S)-5-oxopyrrolidine-2-carboxylate, H-Pyr-OEt, Ethyl (S)-(+)-2-pyrrolidone-5-carboxylate

OR322668-500G
APOSOR322668-500GEA 112 EUR
APOSOR322668-500G
Ethyl-(S)-(+)-2-pyrrolidinone-5-carboxylate 98%
Ethyl-(S)-(+)-2-pyrrolidinone-5-carboxylate
Formel: C₇H₁₁NO₃
Molecular Weight: 157.169 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00064497
CAS-Nummer: 7149-65-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR