5-(tert-Butoxycarbonyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxylic acid

Lieferant: Apollo Scientific

OR322688-100MG
APOSOR322688-100MGEA 40.6 EUR
APOSOR322688-100MG
5-(tert-Butoxycarbonyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxylic acid
5-(tert-Butoxycarbonyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxylic acid
Formel: C₁₂H₁₇N₃O₄
Molecular Weight: 267,281 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10697132
CAS-Nummer: 518990-56-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR