Pyrido[3,4-d]pyrimidine-2,4(1H,3H)-dione 95%

Lieferant: Apollo Scientific

OR322698-1G
APOSOR322698-1GEA 162 EUR
APOSOR322698-1G
Pyrido[3,4-d]pyrimidine-2,4(1H,3H)-dione 95%
Pyrido[3,4-d]pyrimidine-2,4(1H,3H)-dione
Formel: C7H5N3O2
Molecular Weight: 163.1335 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07438024
CAS-Nummer: 21038-67-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR