2-{4-[(6-Chloropyridin-3-yl)methoxy]phenyl}acetic acid, tech

Lieferant: Apollo Scientific

OR32274-1G
APOSOR32274-1GEA 1570 EUR
APOSOR32274-1G
2-{4-[(6-Chloropyridin-3-yl)methoxy]phenyl}acetic acid, tech
2-{4-[(6-Chloropyridin-3-yl)methoxy]phenyl}acetic acid
Formel: C₁₄H₁₂ClNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD05975040
CAS-Nummer: 860784-49-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR