3-Acetyl-4-hydroxy-1-phenyl-1,2-dihydroquinolin-2-one, tech

Lieferant: Apollo Scientific

OR32275-1G
APOSOR32275-1GEA 1570 EUR
APOSOR32275-1G
3-Acetyl-4-hydroxy-1-phenyl-1,2-dihydroquinolin-2-one, tech
3-Acetyl-4-hydroxy-1-phenyl-1,2-dihydroquinolin-2-one
Formel: C₁₇H₁₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00179942
CAS-Nummer: 54289-77-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR