Fmoc-11-aminoundecanoic acid

Lieferant: Apollo Scientific

OR322759-5G
APOSOR322759-5GEA 126 EUR
APOSOR322759-5G
Fmoc-11-aminoundecanoic acid
Fmoc-11-aminoundecanoic acid
Formel: C₂₆H₃₃NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00235891
CAS-Nummer: 88574-07-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR