(E)-N'-[4-Cyano-1-(4-methylphenyl)-1H-pyrazol-5-yl]-N,N-dimethylmethanimidamide, tech

Lieferant: Apollo Scientific

OR32276-1G
APOSOR32276-1GEA 1570 EUR
APOSOR32276-1G
(E)-N'-[4-Cyano-1-(4-methylphenyl)-1H-pyrazol-5-yl]-N,N-dimethylmethanimidamide, tech
(E)-N'-[4-Cyano-1-(4-methylphenyl)-1H-pyrazol-5-yl]-N,N-dimethylmethanimidamide
Formel: C₁₄H₁₅N₅
Molecular Weight: 253,302 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04217411
CAS-Nummer: 78972-84-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR