4-Amino-5-(2-methoxyphenyl)-4H-1,2,4-triazole-3-thiol

Lieferant: Apollo Scientific

OR322764-1G
APOSOR322764-1GEA 183 EUR
APOSOR322764-1G
4-Amino-5-(2-methoxyphenyl)-4H-1,2,4-triazole-3-thiol
4-Amino-5-(2-methoxyphenyl)-4H-1,2,4-triazole-3-thiol
Formel: C₉H₁₀N₄OS
Molecular Weight: 222,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01912738
CAS-Nummer: 61019-26-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR