4-Bromo-2-(4-methylpiperazin-1-yl)benzonitrile

Lieferant: Apollo Scientific

OR322769-500MG OR322769-1G
APOSOR322769-500MGEA 429 EUR
APOSOR322769-500MG APOSOR322769-1G
4-Bromo-2-(4-methylpiperazin-1-yl)benzonitrile
4-Bromo-2-(4-methylpiperazin-1-yl)benzonitrile
Formel: C₁₂H₁₄BrN₃
Molecular Weight: 280.164 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11037544
CAS-Nummer: 1206969-07-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR