4-(4-Methylpiperazino)-1,2-benzenediamine 95%

Lieferant: Apollo Scientific

OR322770-1G OR322770-250MG
APOSOR322770-1GEA 434 EUR
APOSOR322770-1G APOSOR322770-250MG
4-(4-Methylpiperazino)-1,2-benzenediamine 95%
4-(4-Methylpiperazino)-1,2-benzenediamine
Formel: C₁₁H₁₈N₄
Schmelzpunkt: 101…103 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03001209
CAS-Nummer: 54998-08-2
UN: 3259
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR