tert-Butyl 6-chloro-2-iodo-1H-pyrrolo[3,2-c]pyridine-1-carboxylate

Lieferant: Apollo Scientific

OR322773-100MG
APOSOR322773-100MGEA 50.9 EUR
APOSOR322773-100MG
tert-Butyl 6-chloro-2-iodo-1H-pyrrolo[3,2-c]pyridine-1-carboxylate
tert-Butyl 6-chloro-2-iodo-1H-pyrrolo[3,2-c]pyridine-1-carboxylate
Formel: C₁₂H₁₂ClIN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD23378567
CAS-Nummer: 1421933-27-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR