tert-Butyl 2-iodo-1H-imidazole-1-carboxylate

Lieferant: Apollo Scientific

OR322783-250MG
APOSOR322783-250MGEA 841 EUR
APOSOR322783-250MG
tert-Butyl 2-iodo-1H-imidazole-1-carboxylate
tert-Butyl 2-iodo-1H-imidazole-1-carboxylate
Formel: C₈H₁₁IN₂O₂
Molecular Weight: 294,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14585432
CAS-Nummer: 181220-75-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR