5-Iod-2-pyridincarbonitril

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Cyano-5-iodopyridine

OR322789-1G
APOSOR322789-1GEA 317 EUR
APOSOR322789-1G
5-Iod-2-pyridincarbonitril
5-Iod-2-pyridincarbonitril
Formel: C₆H₃IN₂
Molecular Weight: 230,0100 g/mol
Schmelzpunkt: 148…149 °C
Lagertemperatur: Kühlschrank
MDL: MFCD10699132
CAS-Nummer: 41960-47-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR