Methyl 4-cyclopropanecarbonyl-1H-pyrrole-2-carboxylate, tech
Lieferant: Apollo Scientific
OR32279-1G
APOSOR32279-1GEA
1570
EUR
APOSOR32279-1G
Methyl 4-cyclopropanecarbonyl-1H-pyrrole-2-carboxylate, tech
Methyl 4-cyclopropanecarbonyl-1H-pyrrole-2-carboxylate
Formel:
C₁₀H₁₁NO₃ Molecular Weight: 193,199 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07021333 CAS-Nummer: 900019-50-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...