SBAP (Succinimidyl 3-(bromoacetamido)propionate)

Lieferant: Apollo Scientific

Synonyme: SBAP

OR322800-100MG
APOSOR322800-100MGEA 264 EUR
APOSOR322800-100MG
SBAP (Succinimidyl 3-(bromoacetamido)propionate)
SBAP (Succinimidyl 3-(bromoacetamido)propionate)
Formel: C₉H₁₁BrN₂O₅
Molecular Weight: 307,098 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01863457
CAS-Nummer: 57159-62-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR