3-Formyl-4-hydroxybenzoic acid

Lieferant: Apollo Scientific

OR322820-1G
APOSOR322820-1GEA 0 EUR
APOSOR322820-1G
3-Formyl-4-hydroxybenzoic acid
3-Formyl-4-hydroxybenzoic acid
Formel: C₈H₆O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00016595
CAS-Nummer: 584-87-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR