4-Chloro-2-(methylthio)-7H-pyrrolo[2,3-d]pyrimidine

Lieferant: Apollo Scientific

OR322823-250MG
APOSOR322823-250MGEA 105 EUR
APOSOR322823-250MG
4-Chloro-2-(methylthio)-7H-pyrrolo[2,3-d]pyrimidine
4-Chloro-2-(methylthio)-7H-pyrrolo[2,3-d]pyrimidine
Formel: C₇H₆ClN₃S
Molecular Weight: 199.6606 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09025769
CAS-Nummer: 57564-94-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR