(2E,4E)-2-Cyano-N-[(1E)-(dimethylamino)methylidene]-5-phenylpenta-2,4-dienamide, tech

Lieferant: Apollo Scientific

OR32284-1G
APOSOR32284-1GEA 1570 EUR
APOSOR32284-1G
(2E,4E)-2-Cyano-N-[(1E)-(dimethylamino)methylidene]-5-phenylpenta-2,4-dienamide, tech
(2E,4E)-2-Cyano-N-[(1E)-(dimethylamino)methylidene]-5-phenylpenta-2,4-dienamide
Formel: C₁₅H₁₅N₃O
Molecular Weight: 253.299 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00127181
CAS-Nummer: 303995-06-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR