4-(2-Methylpyridin-3-yloxy)-6-chloro-5-Methylpyrimidine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chloro-5-methyl-6-((2-methylpyridin-3-yl)oxy)pyrimidine

OR322841-250MG OR322841-1G
APOSOR322841-250MGEA 92.8 EUR
APOSOR322841-250MG APOSOR322841-1G
4-(2-Methylpyridin-3-yloxy)-6-chloro-5-Methylpyrimidine 97%
4-(2-Methylpyridin-3-yloxy)-6-chloro-5-Methylpyrimidine
Formel: C₁₁H₁₀ClN₃O
Molecular Weight: 235.67 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12756519
CAS-Nummer: 930093-72-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR