Methyl (2Z)-3-(4-chlorophenyl)-2-cyanoprop-2-enoate, tech

Lieferant: Apollo Scientific

OR32285-1G
APOSOR32285-1GEA 1570 EUR
APOSOR32285-1G
Methyl (2Z)-3-(4-chlorophenyl)-2-cyanoprop-2-enoate, tech
Methyl (2Z)-3-(4-chlorophenyl)-2-cyanoprop-2-enoate
Formel: C₁₁H₈ClNO₂
Molecular Weight: 221,64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00158958
CAS-Nummer: 144478-31-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR