Oxazolo[5,4-b]pyridine-2-thiol

Lieferant: Apollo Scientific

OR322851-250MG
APOSOR322851-250MGEA 112 EUR
APOSOR322851-250MG
Oxazolo[5,4-b]pyridine-2-thiol
Oxazolo[5,4-b]pyridine-2-thiol
Formel: C₆H₄N₂OS
Lagertemperatur: Raumtemperatur
MDL: MFCD12755948
CAS-Nummer: 169205-99-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR