Methyl 1-oxo-2-[(pyridin-2-yl)methyl]-1,2-dihydroisoquinoline-4-carboxylate, tech

Lieferant: Apollo Scientific

OR32286-1G
APOSOR32286-1GEA 1570 EUR
APOSOR32286-1G
Methyl 1-oxo-2-[(pyridin-2-yl)methyl]-1,2-dihydroisoquinoline-4-carboxylate, tech
Methyl 1-oxo-2-[(pyridin-2-yl)methyl]-1,2-dihydroisoquinoline-4-carboxylate
Formel: C₁₇H₁₄N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00140339
CAS-Nummer: 303995-32-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR