(2-Cyanophenyl)methanesulphonyl chloride

Lieferant: Apollo Scientific

OR322865-250MG OR322865-1G
APOSOR322865-250MGEA 0 EUR
APOSOR322865-250MG APOSOR322865-1G
(2-Cyanophenyl)methanesulphonyl chloride
(2-Cyanophenyl)methanesulphonyl chloride
Formel: C₈H₆ClNO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD07355846
CAS-Nummer: 51045-34-2
UN: 2923
ADR: 8+6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR