1-Methyl-1H-1,2,3-benzotriazole-5-carbaldehyde

Lieferant: Apollo Scientific

Synonyme: 1-Methyl-1H-benzo[d][1,2,3]triazole-5-carbaldehyde

OR322875-1G
APOSOR322875-1GEA 290 EUR
APOSOR322875-1G
1-Methyl-1H-1,2,3-benzotriazole-5-carbaldehyde
1-Methyl-1H-1,2,3-benzotriazole-5-carbaldehyde
Formel: C₈H₇N₃O
Molecular Weight: 161,161 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03407337
CAS-Nummer: 499770-67-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR