tert-Butyl 3-ethynylphenylcarbamate

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl (3-ethynylphenyl)carbamate

OR322882-1G
APOSOR322882-1GEA 97.3 EUR
APOSOR322882-1G
tert-Butyl 3-ethynylphenylcarbamate
tert-Butyl 3-ethynylphenylcarbamate
Formel: C₁₃H₁₅NO₂
Molecular Weight: 217,264 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14585306
CAS-Nummer: 185619-66-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR