(3E)-1-(4-Bromophenyl)-3-(methoxyimino)propan-1-one, tech

Lieferant: Apollo Scientific

OR32289-1G
APOSOR32289-1GEA 1570 EUR
APOSOR32289-1G
(3E)-1-(4-Bromophenyl)-3-(methoxyimino)propan-1-one, tech
(3E)-1-(4-Bromophenyl)-3-(methoxyimino)propan-1-one
Formel: C₁₀H₁₀BrNO₂
Molecular Weight: 256,096 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00127209
CAS-Nummer: 303995-83-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR