4,7-Dichloro-1H-imidazo[4,5-d]pyridazine 95%

Lieferant: Apollo Scientific
Achtung

OR322895-250MG
APOSOR322895-250MGEA 531 EUR
APOSOR322895-250MG
4,7-Dichloro-1H-imidazo[4,5-d]pyridazine 95%
4,7-Dichloro-1H-imidazo[4,5-d]pyridazine
Formel: C₅H₂Cl₂N₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00723713
CAS-Nummer: 17998-43-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR