4-(3-Tosyl-2-(tosylmethyl)propanoyl)benzoic acid

Lieferant: Apollo Scientific

OR322896-100MG
APOSOR322896-100MGEA 190 EUR
APOSOR322896-100MG
4-(3-Tosyl-2-(tosylmethyl)propanoyl)benzoic acid
4-(3-Tosyl-2-(tosylmethyl)propanoyl)benzoic acid
Formel: C₂₅H₂₄O₇S₂
Molecular Weight: 500.584 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28142486
CAS-Nummer: 124243-00-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR