4-Benzoyl-N,N-dimethyl-1H-pyrrole-2-carboxamide, tech

Lieferant: Apollo Scientific

OR32290-1G
APOSOR32290-1GEA 1570 EUR
APOSOR32290-1G
4-Benzoyl-N,N-dimethyl-1H-pyrrole-2-carboxamide, tech
4-Benzoyl-N,N-dimethyl-1H-pyrrole-2-carboxamide
Formel: C₁₄H₁₄N₂O₂
Molecular Weight: 242,273 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00141841
CAS-Nummer: 303995-89-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR