5-Amino-6-nitroisoindoline-1,3-dione

Lieferant: Apollo Scientific

OR322901-1G
APOSOR322901-1GEA 690 EUR
APOSOR322901-1G
5-Amino-6-nitroisoindoline-1,3-dione
5-Amino-6-nitroisoindoline-1,3-dione
Formel: C₈H₅N₃O₄
Molecular Weight: 207,143 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13186915
CAS-Nummer: 59827-85-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR