(2Z)-1-(4-Chlorophenyl)-3-(dimethylamino)-2-(1H-1,2,4-triazol-1-yl)prop-2-en-1-one, tech

Lieferant: Apollo Scientific

OR32292-1G
APOSOR32292-1GEA 1570 EUR
APOSOR32292-1G
(2Z)-1-(4-Chlorophenyl)-3-(dimethylamino)-2-(1H-1,2,4-triazol-1-yl)prop-2-en-1-one, tech
(2Z)-1-(4-Chlorophenyl)-3-(dimethylamino)-2-(1H-1,2,4-triazol-1-yl)prop-2-en-1-one
Formel: C₁₃H₁₃ClN₄O
Molecular Weight: 276,722 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00215245
CAS-Nummer: 303996-62-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR