1-[1-(1H-Pyrazol-3-yl)ethyl]-1H-1,2,4-triazole 95%

Lieferant: Apollo Scientific

OR32293-1G OR32293-5G
APOSOR32293-1GEA 538 EUR
APOSOR32293-1G APOSOR32293-5G
1-[1-(1H-Pyrazol-3-yl)ethyl]-1H-1,2,4-triazole 95%
1-[1-(1H-Pyrazol-3-yl)ethyl]-1H-1,2,4-triazole
Formel: C₇H₉N₅
Molecular Weight: 163.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00142028
CAS-Nummer: 303996-69-2

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR