6,7-Dihydroxyquinazolin-4(3H)-one

Lieferant: Apollo Scientific

OR322932-100MG
APOSOR322932-100MGEA 224 EUR
APOSOR322932-100MG
6,7-Dihydroxyquinazolin-4(3H)-one
6,7-Dihydroxyquinazolin-4(3H)-one
Formel: C₈H₆N₂O₃
Molecular Weight: 178,145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14583012
CAS-Nummer: 16064-15-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR