4-(Azepan-1-yl)benzaldehyde

Lieferant: Apollo Scientific

OR322936-1G OR322936-500MG
APOSOR322936-1GEA 731 EUR
APOSOR322936-1G APOSOR322936-500MG
4-(Azepan-1-yl)benzaldehyde
4-(Azepan-1-yl)benzaldehyde
Formel: C₁₃H₁₇NO
Molecular Weight: 203.28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06357359
CAS-Nummer: 50333-45-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR