(1E)-1-(Dimethylamino)-4-(1H-1,2,4-triazol-1-yl)pent-1-en-3-one, tech

Lieferant: Apollo Scientific

OR32294-1G
APOSOR32294-1GEA 1570 EUR
APOSOR32294-1G
(1E)-1-(Dimethylamino)-4-(1H-1,2,4-triazol-1-yl)pent-1-en-3-one, tech
(1E)-1-(Dimethylamino)-4-(1H-1,2,4-triazol-1-yl)pent-1-en-3-one
Formel: C₉H₁₄N₄O
Molecular Weight: 194,234 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00664069
CAS-Nummer: 303996-70-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR