4-(2-Chlorophenyl)-2-phenyl-2,3-dihydro-1H-pyrazol-3-one, tech

Lieferant: Apollo Scientific

OR32295-1G
APOSOR32295-1GEA 1570 EUR
APOSOR32295-1G
4-(2-Chlorophenyl)-2-phenyl-2,3-dihydro-1H-pyrazol-3-one, tech
4-(2-Chlorophenyl)-2-phenyl-2,3-dihydro-1H-pyrazol-3-one
Formel: C₁₅H₁₁ClN₂O
Molecular Weight: 270,714 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01316348
CAS-Nummer: 477858-99-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR